Fluke Calibration Thermocouple Adapters for 1523 and 1524 Thermometers
Use to connect 1523 or 1524 reference thermometer to thermocouple leads
  • Supports reference junction compensation (RJC) with its own internal precision thermistor
Keep your thermometers running at their best with these accessories.
ITEM(S) ON SALE
$255.87 - $373.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connections
Availability Pricing
Fluke Calibration 2373-LTC Adapter, Lemo to Universal Thermocouple Probe EW-93202-58 Mfr # 2373-LTC
COMPARE
Lemo to universal thermocouple probe
$373.50USD / Each
$415.00USD / Each
Fluke Calibration 2384-T Smart Connector, Thermocouple Probe (Blue Cap), Spare EW-93202-60 Mfr # 2384-T
COMPARE
$255.87USD / Each
$259.00USD / Each