Fused Silica Capillary Tubing, Deactivated /Pk
Precision Tubing for Performance
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.
$453.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 064050 Fused Silica Capillary Tubing, Deactivated, 363 um OD, 220 um ID, 5 m L; 5/Pk EW-16006-98 Mfr # 064050
COMPARE
$453.00USD / Each
Trajan SGE 064051 Fused Silica Capillary Tubing, Deactivated, 363 um OD, 250 um ID, 5 m L; 5/Pk EW-16006-99 Mfr # 064051
COMPARE
$453.00USD / Each
Trajan SGE 064052 Fused Silica Capillary Tubing, Deactivated, 430 um OD, 320 um ID, 5 m L; 5/Pk EW-16007-00 Mfr # 064052
COMPARE
$453.00USD / Each