Glas-Col Stirring/Heating Mantles

Safely and accurately heat solutions used in various distillations in laboratory and scientific applications
  • Apply heat directly and uniformly to containers, such as round bottom flasks
  • More accurate than hot plates and safer than Bunsen burners
  • Controllers can be purchased individually
Withstanding 400°C (752°F) internal operating temperatures, these mantles provide a smart, accurate way to heat flasks and other containers common within laboratory environments. These motor driven units--which operate from 100 to 1850 rpm--utilize a powerful magnet for higher viscous solutions such as oils.

Available accessories include fabric insulating tops, fabric heating insulating tops, controllers, temperature monitor, and spill guards.
$1,590.00 - $2,040.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Max temperature (° F) Power (VAC)
Availability Pricing
Glas-Col 100D TEM110M Stirring/Heating Mantle, 2L Spherical Flask; 115V EW-36226-75 Mfr # 100D TEM110M
COMPARE
2000 752 115
$1,590.00USD / Each
Glas-Col 100D TEM112M Stirring/Heating Mantle, 3L Spherical Flask; 115V EW-36226-76 Mfr # 100D TEM112M
COMPARE
3000 752 115
$1,600.00USD / Each
Glas-Col 100D TEM114 Stirring/Heating Mantle, 5L Spherical Flask; 115V EW-36226-77 Mfr # 100D TEM114
COMPARE
5000 752 115
$1,705.00USD / Each
Glas-Col 100D TEM116 Stirring/Heating Mantle, 12L Spherical Flask; 115V EW-36226-78 Mfr # 100D TEM116
COMPARE
12000 752 115
$1,895.00USD / Each
Glas-Col 100D TEM118 Stirring/Heating Mantle, 22L Spherical Flask; 115V EW-36226-79 Mfr # 100D TEM118
COMPARE
22000 752 115
$2,040.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat