Global Water Portable, Peristaltic, Fluid Sampling Pumps
Ideal for sampling lakes, ponds, holding pools, or shallow wells
  • Simple control for exact sample size
  • Reversible motors for backflushing of sample hose
  • Rugged, UV-protected PVC case with handle for easy use and transportation
  • Flow rate : up to 500 mL/min at 4-ft head, operating temperature: 0 to 70C
These portable sampling pumps are ideal for sampling lakes, ponds, holding pools, or shallow wells. Samplers are packaged in a lightweight, rugged, UV protected PVC case with handle for easy use and transportation. To protect sample integrity the sample only comes in contact with Norprene and polyethylene tubing which is easily cleaned or replaced between samples.
$1,135.00 - $1,438.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Power
Availability Pricing
Manning MSP250 Fixed Speed, Peristaltic, Fluid Sampling Pump EW-99243-00 Mfr # MS889951-1
COMPARE
Fixed Speed, Peristaltic, Fluid Sampling Pump 12 VDC rechargable battery (included)
$1,135.00USD / Each
Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump EW-99243-05 Mfr # CJ0000
COMPARE
Variable Speed, Peristaltic, Fluid Sampling Pump 12 VDC battery (not included)
$1,438.00USD / Each