Hach HQ4100 Series Portable pH, Conductivity, TDS, Salinity, DO, ORP, ISE Meter, 1 Channel
A robust and intuitive multiparameter meter
  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 100,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
Hach HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), ion selective electrode (ISE), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.
The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 640 x 480 color TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.
$1,836.00 - $2,786.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach HQ4100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel EW-59005-28 Mfr # LEV015.53.4100A
COMPARE
Portable Multi-Meter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel
$1,836.00USD / Each
Hach HQ4100 Portable Multiparameter with Conductivity Electrode, 1 m Cable EW-59005-29 Mfr # LEV015.53.41004
COMPARE
Portable Multi-Meter with Conductivity electrode, 1 m Cable
$2,786.00USD / Each