Hach HQ440d Benchtop LBOD and LDO Multiparameter Meter Kits
Convenience and accuracy of luminescent dissolved oxygen technology
  • IntelliCAL™ smart probes are precalibrated for maximum accuracy
  • Super-bright LCD and large font for easy-to-read results
  • Option to select from 13 languages in setup mode
  • Store up to 500 data sets for GLP-compliant reporting
Versatile HQd meters offer a large graphic display with backlight, and the intuitive user-interface provides guided calibration, setup, and measurement routines to simplify meter operation. Meter has dual-input and multiparameter operation for BOD/DO, pH, ISE, conductivity, and temperature.
Meter can automatically or manually log up to 500 measurements with probe serial number, current calibration data, user ID, sample ID, time, and date. Use the USB port to download data to a PC or flash drive.
The HQd series uses precalibrated Hach IntelliCAL smart probes that store their own calibration data and identification. The meter recognizes each probe, so you can easily switch probes, and the meter is automatically set up to run the next parameter without recalibrating. Kits include laboratory probes with 3-ft (0.9-m) cables.
$5,052.00 - $6,055.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach 8508700 Hq440D Digital pH/Con/LDO Benchtop Meter Kit EW-59004-24 Mfr # 8508700
COMPARE
Hq440D Digital pH/Con/LDO Benchtop Meter Kit
$6,055.00USD / Each
Hach 8508500 HQ440D Benchtop Meter Package with LBOD101 Optical DO Probe EW-59004-26 Mfr # 8508500
COMPARE
HQ440D Benchtop Meter Package with LBOD101 Optical DO Probe
$5,052.00USD / Each