ICI Quick Ship High Performance Fume Hood
High performance fume hood without the need for additional components
  • High performance baffle design for even plenum balance
  • Chemically resistant white poly glass liner
  • Meets SEFA 1 standards
  • Preconfigured to receive 5 service fixtures and alarm
  • Available in 4 ft, 5 ft, and 6 ft widths
ICI Quick Ship fume hoods feature a high-performance design to protect your work. The rear slotted baffle controls pressurization to achieve a safe work environment at 60 FPM without the need for additional components. The high-performance hood features a 24 in (61 cm) deep poly glass lined interior, low static rectangular exhaust duct, a 12 in (30.5 cm) integral stainless steel round duct transition. Hoods are pre-wired for two 120 V 20 amp GFCI duplex receptacles, one vapor tight T8 fluorescent light fixture, one 20 amp light switch, and an outlet for an alarm. It is pre-punched and plugged to receive five service fixtures as well as an optional fan and switch. Meets SEFA1 standards and is UL 1805 listed, ANSI/ASHRAE 110-1995, and UL 1275 compliant.
$13,930.00 - $15,160.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Interior Depth (in) Description
Availability Pricing
ICI Quick Ship High Performance Fume Hood, 48" W EW-78980-92 Mfr # CPF-350-48G7
COMPARE
48 24 ICI Quick Ship High Performance Fume Hood, 48" W
$13,930.00USD / Each
ICI Quick Ship High Performance Fume Hood, 60" W EW-78980-93 Mfr # CPF-350-60G7
COMPARE
60 24 ICI Quick Ship High Performance Fume Hood, 60" W
$15,160.00USD / Each
ICI Quick Ship High Performance Fume Hood, 72" W EW-78980-94 Mfr # CPF-350-72G7
COMPARE
72 24 ICI Quick Ship High Performance Fume Hood, 72" W
$14,625.00USD / Each