Idex® Back Pressure Regulator (BPR) Assemblies
Enhance system performance through outgassing prevention
  • 1/4-28 flat-bottom ports
Select regulators with preset back pressures ranging from 5 to 1000 psi (0.3 to 69 bar). Each assembly comes with a replaceable, factory preset cartridge (except 5 and 20 psi versions).
$133.90 - $183.60USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Housing Material Pressure Setting (PSI)
Availability Pricing
Idex Back Pressure Regulator Assembly, PEEK, 40 psi, 1/16" OD Tubing; 1/EA EW-02014-50 Mfr # P-785
COMPARE
PEEK 40
$136.50USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 75 psi, 1/16" OD Tubing; 1/EA EW-02014-51 Mfr # P-786
COMPARE
PEEK 75
$136.50USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 100 psi, 1/16" OD Tubing EW-02014-52 Mfr # P-787
COMPARE
PEEK 100
$136.50USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 20 psi, 1/16" OD Tubing; 1/EA EW-02014-53 Mfr # P-791
COMPARE
PEEK 20
$136.50USD / Each
Idex Back Pressure Regulator Assembly, SS, 100 psi, 1/16" OD Tubing; 1/EA EW-02014-83 Mfr # U-607
COMPARE
Stainless steel 100
$183.60USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 1000 psi, 1/32" to 1/16" OD Tubing; 1/EA EW-02017-08 Mfr # P-455
COMPARE
PEEK 1000
$136.50USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 250 PSI, 1/16" OD Tubing EW-02017-22 Mfr # P-788
COMPARE
PEEK 250
$133.90USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 500 psi, 1/32" to 1/16" OD Tubing; 1/EA EW-02017-23 Mfr # P-789
COMPARE
PEEK 500
$135.20USD / Each
Idex Back Pressure Regulator Assembly, PEEK, 5 PSI, 1/16" OD Tubing; 1/Ea EW-02017-24 Mfr # P-790
COMPARE
PEEK 5
$135.20USD / Each
Idex Back Pressure Regulator Assembly, Ss, 40 PSI, 1/16" OD Tubing; 1/Ea EW-02017-32 Mfr # U-605
COMPARE
Stainless steel 40
$181.90USD / Each
Idex Back Pressure Regulator Assembly, SS, 75 psi, 1/16" OD Tubing; 1/EA EW-02017-33 Mfr # U-606
COMPARE
Stainless steel 75
$181.90USD / Each
Idex Back Pressure Regulator Assembly, SS, 250 psi, 1/16" OD Tubing; 1/EA EW-02017-34 Mfr # U-608
COMPARE
Stainless steel 250
$181.90USD / Each
Idex Back Pressure Regulator Assembly, SS, 500 psi, 1/16" OD Tubing; 1/EA EW-02017-36 Mfr # U-609
COMPARE
Stainless steel 500
$181.90USD / Each
Idex Back Pressure Regulator Assembly, SS, 750 psi, 1/32" to 1/16" OD Tubing; 1/EA EW-02017-37 Mfr # U-610
COMPARE
Stainless steel 750
$183.60USD / Each
Showing 1- 14 of 14