Idex® Back Pressure Regulator (BPR) Holders
Enhance system performance through outgassing prevention
  • Suitable for use with Idex® back pressure regulator (BPR) cartridges
Choose from multiple holder material options ideal for your application needs.
$128.10 - $336.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Housing Material Max Pressure (PSI) Tubing OD (in)
Availability Pricing
Idex Back Pressure Regulator Holder, PEEK, 1/16" OD Tubing; 1/EA EW-02017-09 Mfr # P-465
COMPARE
PEEK 3000 0.0625
$131.40USD / Each
Idex Back Pressure Regulator Holder, SS, 1/16" OD Tubing; 1/EA EW-02017-29 Mfr # U-469
COMPARE
Stainless steel 4000 0.0625
$128.10USD / Each
Idex Back Pressure Regulator Holder Without Fittings, SS, 10-32 Coned; 1/EA EW-02017-31 Mfr # U-469-02C
COMPARE
Stainless steel
$336.50USD / Each