Idex® Flangeless Headless Fitting Assemblies 1/4-28 Flat-Bottom Connection
Eliminate the need to flange tubing
  • "Headless" head is excellent for tight space applications
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Removable and reusable components saves you time and money. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
$6.00 - $17.70USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Fitting, Headless, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Always in Stock
EW-02020-83 Mfr # XP-283
COMPARE
1/16 PEEK 1/4-28 flat bottom
$6.00USD / Each
Idex Flangeless Fitting, Headless, Natural PPS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA EW-02020-84 Mfr # XP-286
COMPARE
1/16 PPS 1/4-28 flat bottom
$8.00USD / Each
Idex Flangeless Fitting, Headless, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA EW-02021-02 Mfr # XP-348
COMPARE
1/8 PEEK 1/4-28 flat bottom
$17.70USD / Each