Idex® Flangeless Standard Knurl Male Nuts, M6 Flat-Bottom Connection for 1/16" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
$1.60 - $41.10USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA EW-02019-43 Mfr # P-207
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
$1.60USD / Each
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk EW-02019-46 Mfr # P-207X
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk
$15.70USD / Pkg of 10
Idex Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK EW-02019-66 Mfr # P-225YX
COMPARE
Yellow ETFE Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK
$41.10USD / Pkg of 10
Idex Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA EW-02019-67 Mfr # P-226
COMPARE
Black PFA Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
$5.10USD / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PPS, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA EW-02019-81 Mfr # P-288
COMPARE
Natural PPS Flangeless Male Nut, Standard Knurl, Natural PPS, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
$3.40USD / Each