Idex® Flangeless Standard Knurl Male Nuts, M6 Flat-Bottom Connection for 1.8, 2.0, 3.0 mm and 1/8" OD Tubing
Use with multiple size tubing ODs
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Short head available for use in tight-space applications
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Ferrules are available in various materials to suit your application.
$1.60 - $47.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA EW-02019-92 Mfr # P-307
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
$1.60USD / Each
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK EW-02019-93 Mfr # P-307X
COMPARE
Black Acetal Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
$15.70USD / Pkg of 10
Idex Flangeless Male Nut, Short Head, Black PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA EW-02020-39 Mfr # P-347
COMPARE
Black PEEK Flangeless Male Nut, Short Head, Black PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
$3.90USD / Each
Idex Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK EW-02020-41 Mfr # P-347BX
COMPARE
Blue PEEK Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
$47.00USD / Pkg of 10
Idex Flangeless Male Nut, Short Head, Black PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK EW-02020-42 Mfr # P-347X
COMPARE
Black PEEK Flangeless Male Nut, Short Head, Black PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
$38.60USD / Pkg of 10