Idex® High-Pressure Stainless Steel Fitting Assemblies

Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
These 316 stainless steel fitting assemblies include nuts and ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
$99.80 - $136.80USD / Pkg of 10

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (InchShort) Description
Availability Pricing
Idex 316 Stainless Steel Fitting Assembly, Long, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02022-30 Mfr # 6000-211
COMPARE
1/16 10-32 coned 316 Stainless Steel Fitting Assembly, Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
$99.80USD / Pkg of 10
Idex 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02022-31 Mfr # 6000-262
COMPARE
1/16 10-32 coned 316 Stainless Steel Fitting Assembly, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK
$136.80USD / Pkg of 10
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat