Idex® High-Pressure UNF(F) to NPT(M) Threaded Adapters with Fitting
Maintain secure connections to 1/8" and 1/4" NPT ports even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
  • Includes fittings
Made from PEEK, these adapters are durable and have excellent chemical resistance. Wrap NPT threads with thread seal tape to ensure leak-free seal.
$20.40 - $20.80USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Thread size UNF(F) (port 1) (InchShort)
Availability Pricing
Idex Threaded Adapter with Fitting, Red PEEK, 1/16" ID, 1/4-28 Flat Bottom (F) to 1/8" NPT(M) EW-02014-82 Mfr # U-510
COMPARE
Red 1/8 1/4-28 Flat Bottom
$20.60USD / Each
Idex Threaded Adapter with Fitting, Red PEEK, 1/16" ID, 1/4-28 Flat Bottom (F) to 1/4" NPT(M) EW-02024-93 Mfr # U-500
COMPARE
Red 1/8 1/4-28 Flat Bottom
$20.80USD / Each
Idex Threaded Adapter with Fitting, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/4" NPT(M) EW-02024-99 Mfr # U-504
COMPARE
Natural 3/16 5/16-24 Flat Bottom
$20.40USD / Each
Idex Threaded Adapter with Fitting, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/8" NPT(M) EW-02025-03 Mfr # U-514
COMPARE
Natural 3/16 5/16-24 Flat Bottom
$20.40USD / Each