Idex® High-Pressure UNF(F) to UNF Adapters
Maintain secure connections even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.
$18.40 - $43.90USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Thread size UNF(F) (port 2) (InchShort) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Threaded Adapter, PEEK, 0.030" ID, 1/4-28 Flat Bottom (F) to 10-32 Coned (M) EW-02014-11 Mfr # P-652
COMPARE
PEEK 10-32 Coned
$18.40USD / Each
Idex Threaded Adapter, PEEK, 0.040" ID, 10-32 Coned (F) to 1/4-28 Flat Bottom (M) EW-02014-20 Mfr # P-669-01
COMPARE
PEEK 1/4-28 Flat Bottom
$27.70USD / Each
Idex Threaded Adapter, PEEK, 0.125" ID, 5/16-24 Flat Bottom (F) to 1/4-28 Flat Bottom (M) EW-02014-23 Mfr # P-681
COMPARE
PEEK 1/4-28 Flat Bottom
$27.70USD / Each
Idex Threaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F) EW-02024-30 Mfr # P-135
COMPARE
PEEK 5/16-24 Flat Bottom
$19.20USD / Each
Idex Threaded Adapter, PEEK, 0.020" ID, 10-32 Coned (F) to 1/4-28 Flat Bottom (F) EW-02024-37 Mfr # P-627-01
COMPARE
PEEK 1/4-28 Flat Bottom 1/4-28 Flat Bottom
$28.60USD / Each
Idex Threaded Adapter, Extra Long, PEEK, 0.030" ID, 1/4-28 Flat Bottom (F) to 10-32 Coned (M) EW-02024-39 Mfr # P-654
COMPARE
PEEK 10-32 Coned
$21.30USD / Each
Idex Threaded Adapter, PTFE, 0.040" ID, 1/4-28 Flat Bottom (F) to 1/4-28 Flat Bottom (M) EW-02024-42 Mfr # P-671
COMPARE
PEEK 1/4-28 Flat Bottom
$43.90USD / Each
Idex Threaded Adapter, PEEK, 0.050" ID, 1/4-28 Flat Bottom (F) to 10-32 Flat Bottom (M) EW-02024-43 Mfr # P-672
COMPARE
PEEK 10-32 Flat Bottom
$27.60USD / Each
Idex Threaded Adapter, PEEK, 0.040" ID, 5/16-24 Flat Bottom (F) to 1/4-28 Flat Bottom (M) EW-02024-45 Mfr # P-674
COMPARE
PEEK 1/4-28 Flat Bottom
$29.60USD / Each
Idex Threaded Adapter, PEEK, 0.130" ID, 1/2-20 Flat Bottom (F) to 1/4-28 Flat Bottom (M) EW-02024-46 Mfr # P-684
COMPARE
PEEK 1/4-28 Flat Bottom
$34.20USD / Each
Idex Threaded Adapter, PEEK, 0.040" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (M) EW-02024-50 Mfr # P-718
COMPARE
PEEK 5/16-24 Flat Bottom
$28.10USD / Each
Showing 1- 11 of 11