IDEX® High-Purity Plus PFA Tubing
Excellent stress cracking resistance even when exposed to aggressive fluorosurfactants
  • Low gas permeability and outstanding resistance to solvents
  • Manufactured from premium-grade PFA
  • Maximum operating temperature 80ºC
High-Purity Plus PFA tubing withstands repeated flexing and offers high continuous service temperature (recommended max operating temperature 80ºC). Other added benefits include clearer walls, smooth surface texture, and excellent polymer purity.
$34.00 - $321.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (mm) Tubing OD (in)
Availability Pricing
Idex 1902-5 Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.010" ID x 5 ft L; 1/EA EW-02012-79 Mfr # 1902-5
COMPARE
5 0.254 1/16
$34.00USD / Each
Idex 1902L Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.010" ID x 50 ft L; 1/EA EW-02012-80 Mfr # 1902L
COMPARE
50 0.254 1/16
$209.50USD / Each
Idex 1907-5 Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.020" ID x 5 ft L; 1/EA EW-02012-81 Mfr # 1907-5
COMPARE
5 0.5 1/16
$34.00USD / Each
Idex 1907L Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.020" ID x 50 ft L; 1/EA EW-02012-82 Mfr # 1907L
COMPARE
50 0.5 1/16
$207.50USD / Each
Idex 1912-5 Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.030" ID x 5 ft L; 1/EA EW-02012-83 Mfr # 1912-5
COMPARE
5 0.762 1/16
$34.00USD / Each
Idex 1912L Chromatography Tubing, High Purity Plus PFA, 1/16" OD x 0.030" ID x 50 ft L; 1/EA EW-02012-84 Mfr # 1912L
COMPARE
50 0.762 1/16
$207.50USD / Each
Idex 1921-5 Chromatography Tubing, High Purity Plus PFA, 1/8" OD x 0.062" ID x 5 ft L; 1/EA EW-02012-85 Mfr # 1921-5
COMPARE
5 1.575 1/8
$53.60USD / Each
Idex 1921L Chromatography Tubing, High Purity Plus PFA, 1/8" OD x 0.062" ID x 50 ft L; 1/EA EW-02012-86 Mfr # 1921L
COMPARE
50 1.575 1/8
$321.50USD / Each
Showing 1- 8 of 8