Idex® Inline Chromatography Check Valve Accessories

Use with inline check valves
  • Check valve cartridges optimize performance of your check valves
Choose the check valve cartridge suitable for your with your check valve.
$167.30 - $244.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Idex Check Valve Cartridge for 02013-33 and 02015-84, 1.5 psi, 1/16" OD Tubing; 1/EA EW-02015-85 Mfr # CV-3001
COMPARE
Check valves 02013-33 and 02015-84 Check Valve Cartridge for 02013-33 and 02015-84, 1.5 psi, 1/16" OD Tubing; 1/EA
$167.30USD / Each
Idex Check Valve Cartridge for 02015-86, 3 psi, 1/8" OD Tubing; 1/EA EW-02015-87 Mfr # CV-3011
COMPARE
Check valve 02015-86 Check Valve Cartridge for 02015-86, 3 psi, 1/8" OD Tubing; 1/EA
$244.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat