Idex® Low-Pressure Tee Assemblies
Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors. Tee connector assemblies come complete with nuts and ferrules.
ITEM(S) ON SALE
$26.04 - $40.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Fitting Material Orifice Dimensions (mm)
Availability Pricing
Idex Low-Pressure Tee Assembly, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA EW-02008-10 Mfr # P-712
COMPARE
1/16 PEEK 0.5
$40.50USD / Each
Idex Low-Pressure Tee Assembly, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA EW-02008-14 Mfr # P-713
COMPARE
1/8 PEEK 1.25
$40.50USD / Each
Idex Low-Pressure Tee Assembly, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA EW-02014-04 Mfr # P-632
COMPARE
1/16 ETFE 0.5
$26.04USD / Each
$38.70USD / Each
Idex Low-Pressure Tee Assembly, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA EW-02014-05 Mfr # P-633
COMPARE
1/8 ETFE 1.25
$38.70USD / Each
Idex Low-Pressure Tee Assembly, Natural PEEK, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA EW-02014-32 Mfr # P-714
COMPARE
1/16 PEEK 1
$40.00USD / Each