Idex® Low-Pressure Unions with 1/4-28 Flat-Bottom Configuration
Make solid connections in low-pressure applications
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
Use straight and elbow unions in a variety of low-pressure situations throughout your fluidic system. Unions do not include fittings, allowing for customization of fittings. Connect two different size OD tubing by selecting flangeless fittings for 1/16" OD tubing and 1/8" OD tubing.
$3.70 - $82.40USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Orifice Dimensions (mm) Description
Availability Pricing
Idex Low-Pressure Union, True ZDV Female Straight, Natural Acetal, 1/4-28 Flat Bottom; 1/EA EW-02013-94 Mfr # P-603
COMPARE
Acetal Low-Pressure Union, True ZDV Female Straight, Natural Acetal, 1/4-28 Flat Bottom; 1/EA
$3.70USD / Each
Idex Low-Pressure Union, True ZDV Female Straight, Natural Polypropylene, 1/4-28 Flat Bottom; 1/EA
Always in Stock
EW-02013-96 Mfr # P-620
COMPARE
Polypropylene Low-Pressure Union, True ZDV Female Straight, Natural Polypropylene, 1/4-28 Flat Bottom; 1/EA
$3.70USD / Each
Idex Low-Pressure Union, True ZDV Female Straight, Natural ETFE, 1/4-28 Flat Bottom; 1/EA
Always in Stock
EW-02013-97 Mfr # P-623
COMPARE
ETFE Low-Pressure Union, True ZDV Female Straight, Natural ETFE, 1/4-28 Flat Bottom; 1/EA
$6.00USD / Each
Idex Low-Pressure Union, Female Elbow, Natural PEEK, 0.020" Bore, 1/4-28 Flat Bottom; 1/EA EW-02023-19 Mfr # P-430-01
COMPARE
PEEK Low-Pressure Union, Female Elbow, Natural PEEK, 0.020" Bore, 1/4-28 Flat Bottom; 1/EA
$59.40USD / Each
Idex Low-Pressure Union, Female Elbow, Natural PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA EW-02023-21 Mfr # P-432-01
COMPARE
PEEK Low-Pressure Union, Female Elbow, Natural PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
$82.40USD / Each
Idex Low-Pressure Union, Female Straight, Natural PEEK, 0.020" Bore, 1/4-28 Flat Bottom; 1/EA
Always in Stock
EW-02023-70 Mfr # P-702-01
COMPARE
PEEK Low-Pressure Union, Female Straight, Natural PEEK, 0.020" Bore, 1/4-28 Flat Bottom; 1/EA
$15.30USD / Each
Idex Low-Pressure Union, Female Straight, Natural PEEK, 0.050" Bore, 1/4-28 Flat Bottom; 1/EA
Always in Stock
EW-02023-71 Mfr # P-703-01
COMPARE
PEEK Low-Pressure Union, Female Straight, Natural PEEK, 0.050" Bore, 1/4-28 Flat Bottom; 1/EA
$16.30USD / Each
Idex Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA EW-02023-73 Mfr # P-710-01
COMPARE
ETFE Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA
$13.40USD / Each
Showing 1- 8 of 8