Idex® Pressure Relief Valves
Prevent system over pressurization
  • 0
Relief valves protect system components by diverting fluid automatically when inline pressure exceeds the set limit. Use the 100 psi model for general-purpose applications; 5 psi model is perfect for protecting syringe and peristaltic pump systems. To have the pressure relief valve open at a different pressure than 5 or 100 psi, combine a pressure relief tee assembly with a back pressure regulator assembly.
$94.80 - $188.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description Pressure Setting (PSI) Tubing OD (in)
Availability Pricing
Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA EW-02013-95 Mfr # P-612
COMPARE
Idex P-612 Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA 0.0625
$94.80USD / Each
Idex Pressure Relief Valve Assembly, Tee-Shape, 100 psi, 1/16" OD Tubing; 1/EA EW-02014-80 Mfr # U-456
COMPARE
Idex U-456 Pressure Relief Valve Assembly, Tee-Shape, 100 psi, 1/16" OD Tubing; 1/EA 100 0.0625
$188.50USD / Each
Idex Pressure Relief Valve Assembly, Tee-Shape, 5 psi, 1/16" OD Tubing; 1/EA EW-02017-28 Mfr # U-455
COMPARE
Idex Pressure Relief Valve Assembly, Tee-Shape, 5 psi, 1/16" OD Tubing; 1/EA 5 0.0625
$188.50USD / Each