Idex® Standard Knurl Plugs
Available in various thread sizes and materials
  • Easily identify columns with color coded plugs
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
$1.70 - $23.80USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Color Material Compatible With
Availability Pricing
Idex Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA
Always in Stock
EW-02013-77 Mfr # P-309
COMPARE
Black Delrin® acetal 1/4-28 flat-bottom port
$1.70USD / Each
Idex Port Plug, Standard Knurl, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA
Always in Stock
EW-02013-78 Mfr # P-311
COMPARE
Natural ETFE 1/4-28 flat-bottom port
$2.85USD / Each
Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned; 1/EA EW-02013-93 Mfr # P-551
COMPARE
Natural PEEK 10-32 coned port
$3.40USD / Each
Idex Port Plug, Standard Knurl, Black PP, 1/4-28 Flat-Bottom; 1/EA EW-02018-31 Mfr # D-324BLK
COMPARE
Black Polypropylene 1/4-28 flat-bottom port
$2.30USD / Each
Idex Port Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA EW-02018-34 Mfr # P-314
COMPARE
Black ETFE M6 flat-bottom port
$2.60USD / Each
Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread ; 1/EA EW-02018-40 Mfr # P-550
COMPARE
Natural PEEK 10-32 coned port
$3.40USD / Each
Idex Port Plug, Standard Knurl, Natural PEEK, 5/16-24 Flat-Bottom; 1/EA EW-02018-44 Mfr # P-556
COMPARE
Natural PEEK 5/16-24 flat-bottom port
$23.80USD / Each
Idex Port Plug, Standard Knurl, White Acetal, 5/16-24 Flat-Bottom; 1/EA EW-02018-45 Mfr # P-557
COMPARE
White Delrin® acetal 5/16-24 flat-bottom port
$16.20USD / Each
Idex MicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA EW-02024-32 Mfr # P-553
COMPARE
Natural Delrin® acetal 6-32 Coned Port
$7.70USD / Each
Idex MicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA EW-02024-33 Mfr # P-554
COMPARE
White Delrin® acetal 10-32 Coned Port
$8.00USD / Each
Idex High-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA EW-02024-34 Mfr # P-555
COMPARE
Natural PEEK 6-32 Coned Port
$8.50USD / Each
Showing 1- 11 of 11