Idex® Super Flangeless™ Nuts, M6 Flat Bottom
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
$3.40 - $39.10USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-43 Mfr # P-213
COMPARE
1/32 PEEK Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA
$4.10USD / Each
Idex Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK EW-02021-44 Mfr # P-213X
COMPARE
1/32 PEEK Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK
$39.10USD / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-45 Mfr # P-217
COMPARE
1/32 PPS Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA
$3.60USD / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK EW-02021-46 Mfr # P-217X
COMPARE
1/32 PPS Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK
$31.20USD / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-47 Mfr # P-219
COMPARE
1/32 PEEK Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 1/EA
$3.90USD / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK EW-02021-48 Mfr # P-219X
COMPARE
1/32 PEEK Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK
$36.60USD / Pkg of 10
Idex Super Flangeless™ Nut, Headless, Black PPS, 1/16" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-74 Mfr # P-289
COMPARE
1/16 PPS Super Flangeless™ Nut, Headless, Black PPS, 1/16" OD Tubing, M6 Flat Bottom; 1/EA
$4.90USD / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/8" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-75 Mfr # P-317
COMPARE
1/8 PPS Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/8" OD Tubing, M6 Flat Bottom; 1/EA
$3.40USD / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 1/EA EW-02021-76 Mfr # P-319
COMPARE
1/8 PEEK Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 1/EA
$3.90USD / Each
Idex Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 10/PK EW-02021-77 Mfr # P-319X
COMPARE
1/8 PEEK Super Flangeless™ Nut, Standard Knurl, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 10/PK
$36.10USD / Pkg of 10
Idex Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 10/PK EW-02021-90 Mfr # P-337X
COMPARE
1/8 PEEK Super Flangeless™ Nut, Headless, Short, Black PEEK, 1/8" OD Tubing, M6 Flat Bottom; 10/PK
$38.30USD / Pkg of 10
Showing 1- 11 of 11