Idex® Two-Piece Fingertight Fittings with Standard Knurl Head
Popular threaded coned port configurations support quality high-pressure connections
  • Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
$8.00 - $75.80USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 10/PK EW-02013-48 Mfr # F-330X
COMPARE
Natural PEEK 10-32 coned, long thread
$75.80USD / Pkg of 10
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA EW-02019-10 Mfr # F-330
COMPARE
Natural PEEK 10-32 coned, long thread
$8.00USD / Each