Idex® UHMWPE Bottom-of-the-Bottle™ Solvent Filters
Protect your system from damaging particulates
  • Filters are biocompatible
  • Economical—filter cups are replaceable
UHMWPE Filters allow tubing to pass through the bottom of the filter cup to allow solvent to be drawn within 0.10” (2.5 mm) of the bottom of your solvent bottle. Filters are constructed of UHMWPE and ETFE.
$31.70 - $33.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (micron)
Availability Pricing
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA EW-42711-46 Mfr # A-445
COMPARE
1/16" OD tubing 10
$33.80USD / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPE/ETFE, 10 µm, 1/8" OD Tubing; 1/EA EW-42711-47 Mfr # A-446
COMPARE
1/8" OD tubing 10
$31.70USD / Each