Idex® VHP MicroTight® Unions
Maintain secure connections even under high pressure
  • 316 stainless steel body withstands high pressures Holds tight connection even through vibration
VHP MicroTight® Unions join two pieces of tubing in a connection that will withstand higher pressures using trademarked MicroTight technology.
$97.20 - $165.90USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Operating Pressure (PSI) Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex VHP MicroTight Union Assembly, PEEK/SS, 0.006" ID, 1/32" OD Tubing, 5/16-24 Coned; 1/EA EW-02014-84 Mfr # UH-432
COMPARE
15000 1/32 SS/PEEK
$165.90USD / Each
Idex VHP MicroTight Union Assembly, PEEK/SS, 0.006" ID, 360 µm OD Tubing, 5/16-24 Coned; 1/EA EW-02025-08 Mfr # UH-436
COMPARE
15000 1/64 SS/PEEK
$124.50USD / Each
Idex VHP MicroTight Union, True ZDV, PEEK/SS, 1/32" OD Tubing, 6-32 Coned; 1/EA EW-02025-11 Mfr # UH-632
COMPARE
15000 1/32 SS/PEEK
$97.20USD / Each