Idex® VHP (Very High Pressure) Chromatography Micro Fittings
Fittings for very high pressure applications
  • Ideal for UHPLC applications Quick component replacement with minimal down time
  • Max pressure of 15,000 psi (1035 bar)
  • Requires 4.0 in-lb (0.45 N-m) of torque
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. This patented line of VHP fittings are designed to work in extreme pressure environments. They are perfect for use in high-performance analytical systems. Use with microferrules made from PK (a proprietary PEEK blend) that allows them to be used at higher pressures and temperatures (up to 200° C).
$8.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Thread size UNF(F) (port 1) (InchShort)
Availability Pricing
Idex VHP MicroTight® Female Nut, Natural PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA EW-02022-53 Mfr # P-416
COMPARE
Natural 5/16-24 coned
$8.00USD / Each
Idex VHP MicroTight® Female Nut, Black PEEK, 1/32" OD Tubing, 5/16-24 Coned; 1/EA EW-02022-54 Mfr # P-416BLK
COMPARE
Black 5/16-24 coned
$8.00USD / Each