IKA® RC 5 Series Recirculating Chillers
Powerful, eco-friendly recirculating chiller designed for applications down to –30°C
  • Uses environmentally friendly natural refrigerant R290
  • Large display for easy setting and viewing of parameters
  • RS-232 and USB interfaces—connect to a PC to control the recirculating chillers
  • Feed hopper and bleed valve offer safe and clean handling of the thermofluid
  • Easy access air filter makes cleaning convenient
Use RC 5 recirculating chillers for fast and efficient cooling of external devices such as rotary evaporators, reflux condensers, and calorimeters. Chillers come with an environmentally friendly natural refrigerant R290 which significantly increases the efficiency of the chiller while meeting EPA guidelines.
PID temperature control provides excellent accuracy and stability to handle most demanding applications. Connect the chiller to a PC via the USB or RS-232 interface; control and monitor with the labworldsoft® laboratory software. A built-in variable-speed PEEK pump allows continuous adjustment of maximum pressure and flow rate. Fluid level indicator turns LED segment red if the fluid level is too high or low.
$9,668.00 - $11,372.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Reservoir Capacity (L) Temperature Stability (° C) Description
Availability Pricing
IKA RC 5 Basic Recirculating Chiller, –30 to 20°C; 115V EW-28711-75 Mfr # 20008503
COMPARE
7 ±0.2 RC 5 basic Recirculating Chiller, –30 to 20°C; 115V
$9,668.00USD / Each
IKA RC 5 Control Recirculating Chiller with Wireless Controller, –30 to 20°C; 115V EW-28711-76 Mfr # 20008504
COMPARE
7 ±0.1 RC 5 control Recirculating Chiller with Wireless Controller, –30 to 20°C; 115V
$11,372.00USD / Each