Julabo MAGIO™ Circulator Accessories
For use with the MAGIO™ range of refrigerated, heated, and immersion circulators/baths
  • Optimize performance of your refrigerated/heating circulators with these accessories
Choose from accessories compatible with the MAGIO™ range of circulators include an electronic module to allow for analog connections to the pump, a condensation trap with lid, and lockable lid. Both bath lid accessories increase energy efficiency by preventing steam leakage and the associated fluid loss.
$953.00 - $2,949.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Julabo Electronic Module with Analog Connections forMAGIO MS and MX Units EW-12153-10 Mfr # 9900100
COMPARE
Julabo MAGIO MS and MX Refrigerated/Heating Circulators Electronic Module with Analog Connections forMAGIO MS and MX Units
$953.00USD / Each
Julabo Condensation Trap with Bath Lid for 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators EW-12153-11 Mfr # 9970700
COMPARE
Julabo MAGIO MS, CORIO CP and CD, DYNEO DD models 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators Condensation Trap with Bath Lid for 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators
$2,949.00USD / Each
Julabo Lockable Bath Lid for 600F, 601F, 1000F, 1000FW, and BC12 Refrigerated/Heating Circulators EW-12153-74 Mfr # 9970243
COMPARE
Julabo MAGIO, CORIO CP and CD, DYNEO DD models BC12, 600F, 601F, 1000F, and 1000FW Refrigerated/Heating Circulators Lockable Bath Lid for 600F, 601F, 1000F, 1000FW, and BC12 Refrigerated/Heating Circulators
$2,352.00USD / Each