Koflo Disposable Static Mixers
Ideal for mixing short-life fluids through a pipe line
  • Able to mix low to high viscosity components
  • Maximum operating temperature is 170 °F (76.7 °C)
  • Chemical resistant
These economical unmounted static mixers are ideal for in-line mixing of short-life fluids such as adhesives, epoxies, or coatings. The mixers feature helical-style mixing elements which allow for pressure drop-efficient mixing of low, medium, and high viscosity fluids across all flow regimes.
Chemically resistant polyacetal elements can be used with any standard tubing having the same tubing ID as the element OD. Available in several element configurations and sizes to meet your small-scale mixing requirements.
$4.50 - $6.30USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Element OD (in) Mixer Length (in) No. Of Elements
Availability Pricing
Koflo P-.25-12 Disposable Static Mixer, 1/4" dia, 12 Element
Always in Stock
EW-14669-00 Mfr # P-.25-12
COMPARE
0.25 3.1 12
$4.50USD / Each
Koflo P-.375-12 Disposable Static Mixer, 3/8" Dia, 12 ELEMENT
Always in Stock
EW-14669-01 Mfr # P-.375-12
COMPARE
0.375 4.6 12
$5.30USD / Each
Koflo P-.5-12 Disposable Static Mixer,,  1/2" Dia, 12 ELEMENT
Always in Stock
EW-14669-02 Mfr # P-.5-12
COMPARE
0.5 6.1 12
$6.30USD / Each