Koflo Polypropylene Tube Disposable Static Mixers, Plain Ends
Ideal for mixing short-life fluids through a pipe line
  • Able to mix low to high viscosity components
  • Accepts compression tube fittings with same tubing OD
  • Maximum operating temperature is 170 °F (76.7 °C)
  • Chemical resistant
These economical static mixers are ideal for in-line mixing of short-life fluids such as adhesives, epoxies, or coatings. The mixers feature helical-style mixing elements which allow for pressure drop-efficient mixing of low, medium, and high viscosity fluids across all flow regimes.
Chemically resistant polyacetal elements and polypropylene tubing with plan ends resist many chemical solutions. Available in several element configurations and sizes to meet your small-scale mixing requirements.
$8.00 - $15.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Mixer Length (in) Tubing ID (in) Tubing OD (in)
Availability Pricing
Koflo PPT-.25-12-P Polypropylene Tube Disposable Static Mixers, 1/4" OD (1/8" ID) , 12 Element
Always in Stock
EW-14669-03 Mfr # PPT-.25-12-P
COMPARE
2.25 0.125 0.25
$8.00USD / Each
Koflo PPT-.25-24-P Polypropylene Tube Disposable Static Mixers, 1/4" OD (1/8" ID), 24 Element
Always in Stock
EW-14669-04 Mfr # PPT-.25-24-P
COMPARE
3.75 0.125 0.25
$11.50USD / Each
Koflo PPT-.375-12-P Polypropylene Tube Disposable Static Mixers, 3/8" OD (1/4" ID), 12 Element EW-14669-05 Mfr # PPT-.375-12-P
COMPARE
3.75 0.25 0.375
$9.00USD / Each
Koflo PPT-.375-24-P Polypropylene Tube Disposable Static Mixers, 3/8" OD (1/4" ID), 24 Element EW-14669-06 Mfr # PPT-.375-24-P
COMPARE
6.75 0.25 0.375
$11.50USD / Each
Koflo PPT-.5-12-P Polypropylene Tube Disposable Static Mixers, 1/2" OD (3/8" ID), 12 Element EW-14669-07 Mfr # PPT-.5-12-P
COMPARE
5.5 0.375 0.5
$9.50USD / Each
Koflo PPT-.5-24-P Polypropylene Tube Disposable Static Mixers, 1/2" OD (3/8 " ID), 24 Element EW-14669-08 Mfr # PPT-.5-24-P
COMPARE
10.5 0.375 0.5
$12.00USD / Each
Koflo PPT-.625-12-P Polypropylene Tube Disposable Static Mixers, 5/8" OD (1/2" ID), 12 Element EW-14669-09 Mfr # PPT-.625-12-P
COMPARE
7.25 0.5 0.625
$13.50USD / Each
Koflo PPT-.625-24-P Polypropylene Tube Disposable Static Mixers, 5/8" OD (1/2" ID), 24 Element EW-14669-10 Mfr # PPT-.625-24-P
COMPARE
13.5 0.5 0.625
$15.00USD / Each
Showing 1- 8 of 8