LabChem Mercuric Nitrate for Chloride, 0.141 N
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$112.25 - $403.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Mercuric Nitrate for Chloride, 0.141 N (0.071 M); 500 mL EW-80028-21 Mfr # LC166601
COMPARE
500 Mercuric Nitrate, 0.141 N (0.071 M) (1 mL=5 mg Clˉ), for Chloride; 500 mL
$112.25USD / Each
LabChem Mercuric Nitrate for Chloride, 0.141 N (0.071 M); 1 L EW-80028-22 Mfr # LC166602
COMPARE
1000 Mercuric Nitrate, 0.141 N (0.071 M) (1 mL=5 mg Clˉ), for Chloride; 1 L
$172.25USD / Each
LabChem Mercuric Nitrate for Chloride, 0.141 N (0.071 M); 4 L EW-80028-27 Mfr # LC166604
COMPARE
4000 Mercuric Nitrate, 0.141 N (0.071 M) (1 mL=5 mg Clˉ), for Chloride; 4 L
$403.00USD / Each