LabChem Mercuric Thiocyanate, 0.42% in Methanol
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$77.50 - $120.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride; 500 mL EW-80028-34 Mfr # LC167201
COMPARE
500 Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride; 500 mL
$77.50USD / Each
LabChem Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride; 1 L EW-80028-35 Mfr # LC167202
COMPARE
1000 Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride; 1 L
$120.50USD / Each