LaMotte Dissolved Oxygen Test Kit
Color titration method determines dissolved oxygen levels
  • Winkler titration method to test samples or calibrate meters
  • Inexpensive backup in case of meter failure
This test is an azide modification of the Winkler method where oxygen, in the presence of a strong alkali, oxidizes manganese, which in turn reacts with iodide to form iodine. This is titrated with a standard thiosulfate solution in the presence of a starch indicator to enhance the endpoint. Azide eliminates the nitrite interference. Test kit and refills contain enough reagents for 50 tests at an oxygen level of 10 ppm.
$109.10USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Gallons Of Water For 1 ppm (Gal) Measurement Points Test
Availability Pricing
LaMotte 5860 Dissolved Oxygen Test Kit, 50 tests/kit EW-53003-00 Mfr # 5860-01
COMPARE
0 to 10 ppm Dissolved oxygen
$109.10USD / Each