LaMotte® Soil Test Kits
Quick, visible color-change indicates results
  • Refillable reagents minimize product usage costs
Soil test kits simplify measuring the pH level, nutrients, or texture of your soil. This line of combination soil analysis outfits offers the finest visual color matching system available to today’s agronomist. Technically advanced reagent systems and unique extraction procedures based on the Mehlich I extraction provide fast, simple and extremely accurate soil testing. A series of rapid, accurate chemical tests use standardized reagents to produce color reactions measured against laminated color charts.
Tests for calcium, chloride, potassium, and sulfate are based on turbidity measurements. Refill reagents are available to replenish your kit.
$131.80 - $1,119.30USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
LaMotte 1067 Soil Texture Test Kit for Sand, Silt, and Clay Fractions; 50 Tests EW-99030-00 Mfr # 1067
COMPARE
Soil Texture Test Kit for Sand, Silt, and Clay Fractions; 50 Tests
$131.80USD / Each
LaMotte 5010-01 Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests EW-99031-10 Mfr # 5010-01
COMPARE
Soil Nutrient Test Kit for Macronutrient, Micronutrient, and pH; 50 Tests
$1,119.30USD / Each