MadgeTech Pressure - Temperature Data Loggers
The MH1000PR series is a battery powered, stand-alone pressure and temperature data logger. This all-in-one compact, portable, easy to use device measures and records up to 16,383 pressure and 16.383 temperature measurements. The data logger accommodates numerous adaptors for various pressure ranges and pipefittings. Its real time clock ensures that all data is time and date stamped. The storage medium is non-volatile solid-state memory, providing maximum data security even if the battery becomes discharged. The device can be started and stopped directly from your computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software does the rest.
FEATURES
  • Rugged, stainless steel case
  • Submersible
  • Programmable start time
  • Real-time operation
  • User-friendly
  • Low cost
  • ¼" NPT fitting
  • Readings selectable 1 every 2 sec to 1 every 12 hour
$837.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech PRTEMP1000-30-PSIG Temp/Pressure Recorder; 30 psig, 1/4"NPT EW-18003-33 Mfr # PRTEMP1000-30-PSIG
COMPARE
176
$837.00USD / Each