MAPA Temp-Tec Heat-Insulated Neoprene Gloves
Two-tone wear indicator warns you when to replace gloves
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
These gloves are made of synthetic neoprene rubber for good chemical resistance. Easily maintain your grip with the nonslip, roughened surface. Withstand temperatures from -100 to 480°F (-73 to 249°C). The 14"L gloves feature integral cotton fleece liner.
$50.60USD / Pair of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
MAPA 332428 Heat-Insulated Neoprene Gloves, Medium
Buy More Save More
EW-40211-30 Mfr # 332428
COMPARE
14 Medium
$50.60USD / Pair of 1
MAPA 332429 Heat-Insulated Neoprene Gloves, Large
Buy More Save More
EW-40211-31 Mfr # 332429
COMPARE
14 Large
$50.60USD / Pair of 1
MAPA 332420 Heat-Insulated Neoprene Gloves, Extra-large
Buy More Save More
EW-40211-32 Mfr # 332420
COMPARE
14 X-Large
$50.60USD / Pair of 1