Marsh Bellofram Type 70 Medium-Flow Regulators
Flow capacity to 80 scfm!
  • Responds to minute changes in pressure
  • Diaphragm constructed of Buna-N and polyester- designed for millions of cycles
These Type 70 regulators are specifically designed for applications that require substantial flow capacity and accurate pressure controls. Downstream pressure can be set within 0.25" of water column and is accurately maintained under varying flow conditions with the help of an aspirator tube, which adjusts the air supply in accordance with the flow velocity. A balanced supply valve, using a rolling diaphragm, makes the regulator virtually immune to changes in supply pressure. Quality materials ensure long, trouble-free operation in the most difficult industrial environments. The precision die-cast aluminum housing is finished with vinyl paint, which resists scratching, weathering, and other physical wear. Regulators are pressure-tested and chrome-plated for corrosion resistance.
$102.50 - $257.25USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Control Range (high) (PSI) Control Range (low) (PSI)
Availability Pricing
Marsh Bellofram 960-094-000 2-150 Psig Precision High Flow Air Regulator Pipe Panel Or Bracket Mount EW-68458-47 Mfr # 960-094-000
COMPARE
150 2
$218.25USD / Each
Marsh Bellofram 960-177-000 Filter Regulators, 0 to 100 psig, 40 um EW-68458-51 Mfr # 960-177-000
COMPARE
100 0
$102.50USD / Each
Marsh Bellofram 960-129-000 High- Flow Regulators, 1/4" NPT(F), 0 to 2 psi, 40 scfm EW-68825-36 Mfr # 960-129-000
COMPARE
2 0
$175.25USD / Each
Marsh Bellofram 960-130-000 High- Flow Regulators, 1/4" NPT(F), 0 to 10 psi, 40 scfm EW-68825-38 Mfr # 960-130-000
COMPARE
10 0
$164.00USD / Each
Marsh Bellofram 960-089-000 High- Flow Regulators, 3/8" NPT(F), 0 to 30 psi, 40 scfm EW-68825-40 Mfr # 960-089-000
COMPARE
30 0
$164.00USD / Each
Marsh Bellofram 960-092-000 High- Flow Regulators, 1/4" NPT(F), 1 to 60 psi, 40 scfm EW-68825-42 Mfr # 960-092-000
COMPARE
60 1
$164.00USD / Each
Marsh Bellofram 960-152-000 High-Flow Regulator, 3 to 200 psi, 40 scfm EW-68825-46 Mfr # 960-152-000
COMPARE
200 3
$223.00USD / Each
Bellofram 960-174-000 High- Flow Regulators, 3/8" NPT(F), 0 to 2 psi, 50 scfm EW-68825-48 Mfr # 960-174-000
COMPARE
2 0
$175.25USD / Each
Bellofram 960-131-000 High- Flow Regulators, 3/8" NPT(F), 0 to 10 psi, 50 scfm EW-68825-50 Mfr # 960-131-000
COMPARE
10 0
$164.00USD / Each
Marsh Bellofram 960-090-000 High- Flow Regulators, 1/4" NPT(F), 0 to 30 psi, 50 scfm EW-68825-52 Mfr # 960-090-000
COMPARE
30 0
$218.25USD / Each
Marsh Bellofram 960-093-000 High-Flow Regulator, 2 to 150 psi, 50 scfm EW-68825-56 Mfr # 960-093-000
COMPARE
150 2
$164.00USD / Each
Bellofram 960-162-000 High- Flow Regulators, 1/2" NPT(F), 0 to 2 psi, 80 scfm EW-68825-60 Mfr # 960-162-000
COMPARE
2 0
$200.25USD / Each
Marsh Bellofram 960-163-000 High- Flow Regulators, 1/2" NPT(F), 0 to 10 psi, 80 scfm EW-68825-62 Mfr # 960-163-000
COMPARE
10 0
$200.25USD / Each
Bellofram 960-159-000 High- Flow Regulators, 1/2" NPT(F), 0 to 30 psi, 80 scfm EW-68825-64 Mfr # 960-159-000
COMPARE
30 0
$200.25USD / Each
Marsh Bellofram 960-161-000 High-Flow Regulator, 2 to 150 psi, 80 scfm EW-68825-68 Mfr # 960-161-000
COMPARE
150 2
$200.25USD / Each
Marsh Bellofram 960-164-000 High-Flow Regulator, 3 to 200 psi, 80 scfm EW-68825-70 Mfr # 960-164-000
COMPARE
200 3
$257.25USD / Each
Showing 1- 16 of 16