Mettler Toledo Cuvettes for EasyPlus Visible and UV/Visible Spectrophotometers
Great optical transparency of >80% with excellent chemical stability
  • Designed for long-term use, making them environmentally friendly and economical
  • Comes in a matching pair
These macro cuvettes offer reliability and accuracy at an economical cost. Small, clear rectangular cuvettes feature >80% optical transparency, ensuring accurate results. Reusable glass cuvettes are environmentally friendly and provide an economical alternative to disposable cuvettes.
$29.20 - $291.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (MicroL) Pathlength (mm)
Availability Pricing
Mettler Toledo 30675051 Cuvette for Easy UV UV/Visible Spectrophotometers, Standard Q Quartz Glass, 10 mm
New Products
EW-33332-33 Mfr # 30675051
COMPARE
Quartz glass 3500 10
$146.00USD / Each
Mettler Toledo 30675052 Cuvette for Easy UV UV/Visible Spectrophotometers, Standard Q Quartz Glass, 50 mm
New Products
EW-33332-34 Mfr # 30675052
COMPARE
Quartz glass 17500 50
$291.00USD / Each
Mettler Toledo 30675053 Cuvette for Easy VIS Visible Spectrophotometers, Standard O Optical Glass, 10 mm
New Products
EW-33332-35 Mfr # 30675053
COMPARE
Optical glass 3500 10
$29.20USD / Each
Mettler Toledo 30675054 Cuvette for Easy VIS Visible Spectrophotometers, Standard O Optical Glass, 50 mm
New Products
EW-33332-36 Mfr # 30675054
COMPARE
Optical glass 17500 50
$58.10USD / Each