Mettler Toledo EasyPlus Visible and UV/Visible Spectrophotometers with ScanStart™ Bundle
Analyze Macherey-Nagel’s NANOCOLOR® test kits with predefined and verified methods
  • Perform conventional spectrophotometry, color measurements, and water analyses using one instrument
  • 3-in-1 functionality saves bench space and reduces your investment in equipment and service
  • ScanStart™ bundle with a handheld barcode scanner offers easy water testing workflow for NANOCOLOR® water test kits
  • Fast and accurate—photometric data automatically calculated using preprogrammed methods
  • PathDetect™ feature checks the path length ensuring accurate results
  • Quick setup using user-friendly 4.3” touchscreen and interface with workflow guidance and integrated tutorials
  • SmartLid™ feature automatically starts the measurement when the lid closes, increasing productivity
  • Washable drip tray for fast and easy clean up
The Mettler Toledo EasyPlus ScanStart™ spectrophotometers combine conventional spectrophotometry, color measurements, water analyses, plus the ability to analyze Macherey-Nagel’s NANOCOLOR® test kits in one instrument. The ScanStart bundle comes with a handheld barcode scanner to facilitate the workflow. Simply download the method package from mt.com. All methods contain predefined barcode content, ensuring accuracy and efficiency.
Easily access LongClick™ feature to start the last performed method and a favorite list to eliminate calculation and transcription errors. Automatic calculation of photometric data using preprogrammed methods provides quick results. Methods that are compliant to standards such as ASTM, DIN, ISO, and EPA eliminate the need for post-measurement calculation and data transcription to achieve accurate results efficiently.
Color measurements are obtained from absorbance or transmittance measurements converted to a color value according to the selected color scale. Enjoy fast, compliant color measurements—up to 30 color scales are available including CIELAB, APHA, Gardner, and more. Order SQKitReader™ barcode reader 83700-80 to read the 2D barcode of Spectroquant® water test kits for access to over 150 Spectroquant water test methods.
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Min Wavelength (nm) Source Lamp
Availability Pricing
Mettler Toledo 30997809 Easy UV UV/Visible Spectrophotometer ScanStart™ Bundle with Handheld Scanner EW-33332-31 Mfr # 30997809
COMPARE
3 nm 190 Xenon flash lamp
$6,955.00USD / Each
Mettler Toledo 30997810 Easy VIS Visible Spectrophotometer ScanStart™ Bundle with Handheld Scanner EW-33332-32 Mfr # 30997810
COMPARE
4 nm 330 Tungsten lamp
$4,710.00USD / Each