Mettler Toledo LA Series Affordable Quality Toploading Balances
Quality balances offer accurate and reliable results at a low cost
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Easy-to-read, bright backlit LCD
  • Connect to a printer, secondary display, or computer via the RS-232 interface
  • Make adjustments with external weights to ensure accuracy
  • Overload protection guards the weighing cell against excess weight overload
  • Durable die-cast aluminum base and ABS top housing
The LA series toploading balances provide reliable performance at a low cost. Balances are simple to use yet feature three built-in applications to perform calculations automatically, ensuring consistent processes and fewer errors. Perform weighing operations below the balance with the included weighing hook.
Rugged housing, made of high-quality and durable materials, protects the sensitive sensor from environmental influences and impacts. Setting protection feature protects balance configurations against unintended changes. Built-in overload protection prolongs the life of your balance.
$1,355.00 - $1,795.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo LA403E Precision Toploading Balance, LA Series, 420g x 1mg with LCD Display and External Calibration EW-02629-64 Mfr # 30823722
COMPARE
420 1 Precision Toploading Balance, 420 g x 1 mg with Draft Shield
$1,525.00USD / Each
Mettler Toledo LA4002E Precision Toploading Balance, LA Series, 4200g x 10mg with LCD Display and External Calibration EW-02629-65 Mfr # 30823723
COMPARE
4200 10 Precision Toploading Balance, 4200 g x 10 mg
$1,795.00USD / Each
Mettler Toledo LA203E Precision Toploading Balance, LA Series, 220g x 1mg with LCD Display and External Calibration EW-02629-66 Mfr # 30853154
COMPARE
220 1 Precision Toploading Balance, 220 g x 1 mg with Draft Shield
$1,355.00USD / Each
Mettler Toledo LA2002E Precision Toploading Balance, LA Series, 2200g x 10mg with LCD Display and External Calibration EW-02629-67 Mfr # 30853155
COMPARE
2200 10 Precision Toploading Balance, 2200 g x 10 mg
$1,475.00USD / Each