Mettler Toledo NewClassic Moisture Balances
Fast, precise, and reliable moisture determination
  • Easy three-step process
  • Results in just a few minutes
  • Quick set-up
  • Robust design
  • RS-232 interface connects to external printer
Designed for laboratory and production applications for determining the moisture content of products to maintain quality and ensure optimum shelf-life. When testing is complete, unit cools down quickly for greater sample throughput and user safety. RS-232 interface connects analyzer to an external printer.
ITEM(S) ON SALE
$3,985.80 - $7,098.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
Mettler Toledo HE53 (115V) Newclassic He Halogen Moisture Balance, 54G x 1mg EW-11335-01 Mfr # 30100251
COMPARE
54 Halogen 0.01
$3,985.80USD / Each
$4,380.00USD / Each
Mettler Toledo HE73 (115V) Newclassic He Halogen Moisture Balance, 71g x 1mg EW-11335-03 Mfr # 30237205
COMPARE
71 Halogen 0.001
$5,330.00USD / Each
Mettler Toledo HC103 NewClassic HC Halogen Moisture Balance, 101 g x 0.1% EW-11335-05 Mfr # 30216103
COMPARE
101 Halogen 0.001
$7,098.00USD / Each
$7,800.00USD / Each