Microflex Diamond Grip Plus Latex Exam Gloves
Thick design provides durable protection
  • Medical examination grade protects hands of healthcare professionals
  • Fully textured surface offers secure grip
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
These exam gloves feature a palm thickness of 5.1 mil and a finger thickness of 6.3 mil for durable protection. Gloves feature a fully textured surface for excellent grip — wet or dry. Natural latex ensures superior comfort and resilience. Beaded cuffs improve tear resistance and protect hands.
Ideal for use in the food processing, life sciences, healthcare, utilities, and warehousing industries.
Standards and certifications include ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
$17.60 - $19.60USD / Box of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Palm Thickness (mil) Size
Availability Pricing
Microflex DGP350-S Diamond Grip Plus Latex Gloves; Small
Buy More Save More
EW-86232-30 Mfr # DGP350-S
COMPARE
9 1/2 5.1 Small
$19.60USD / Box of 100
Microflex DGP350-M Diamond Grip Plus Latex Gloves; Medium
Buy More Save More
EW-86232-31 Mfr # DGP350-M
COMPARE
9 1/2 5.1 Medium
$17.60USD / Box of 100
Microflex DGP350-L Diamond Grip Plus Latex Gloves; Large
Buy More Save More
EW-86232-32 Mfr # DGP350-L
COMPARE
9 1/2 5.1 Large
$17.60USD / Box of 100
Microflex DGP350-XL Diamond Grip Plus Latex Gloves; Extra-Large
Buy More Save More
EW-86232-33 Mfr # DGP350-XL
COMPARE
9 1/2 5.1 X-Large
$17.60USD / Box of 100