MLA Adjustable- and Selectable-Volume Pipettors
Calibration feature is handy for viscous solutions
These pipettes feature special benefits without compromising MLA quality. Built with aluminum alloy and stainless steel, the lifetime warranty ensures maximum durability and performance. Pipets are engraved with serial numbers for easy traceability and quality control. Adjust or calibrate pipettors above or below the stated volume up to ±10%.
$496.00 - $738.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Max Volume (mL) Min Volume (mL)
Availability Pricing
VistaLab 1061 Adjustable Volume Pipette, 1 to 5 mL; Each EW-06199-13 Mfr # 1061
COMPARE
±1.00% 5 1
$680.00USD / Each
VistaLab MLA Volume Pipette, 50/100/200 uL, Silver; Each EW-06199-35 Mfr # 1132
COMPARE
±1.30% at 200.0µL; ±1.50% at 100.0µL; ±2.50% at 50.00µL 0.2 0.05
$496.00USD / Each
VistaLab 1134 Volume Pipette, 2/3/4/5 mL, Blue; Each EW-06199-37 Mfr # 1134
COMPARE
±1.00% 5 2
$738.00USD / Each