MTC Bio Laboratory Label Printer Cassettes
Ideal label identification for a variety of laboratory applications
  • Can be autoclaved, boiled, or frozen
  • Manufacturer defect warranty
These label printer cassette tapes are ideal for label identification for nearly any laboratory application. Compatible with MTC Bio handheld and Bluetooth® enable laboratory label printers, these tapes can be autoclaved, boiled or frozen to temperatures as low as -50 °C.
Choose from 13 label cassettes tapes available in a variety of colors, including transparent. Three label heights are available for different sized vessels. The 6 mm tapes are great for tissue culture plates, 12 mm tapes are ideal from microtubes, and the extra tall 24 mm tapes are ideal for large tubes and flasks.
Special "ultra-low" label tapes (models 06530-57 & 06530-62) are available for storage in -80 °C environments, or for liquid nitrogen applications.
$32.40 - $53.20USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Color Length (InchShort)
Availability Pricing
MTC Bio 26' Cassette of 12mm Lab Tape, Blue w/ White Print
Buy More Save More
EW-06530-53 Mfr # L9010-12BW
COMPARE
Blue with white print 312
$32.40USD / Each
MTC Bio 26' Cassette of 12mm Lab Tape, clear w/ black Print
Buy More Save More
EW-06530-54 Mfr # L9010-12CK
COMPARE
Clear with black print 312
$41.50USD / Each
MTC Bio   26' Cassette of 12mm Lab Tape, Green w/ White Print
Buy More Save More
EW-06530-55 Mfr # L9010-12GW
COMPARE
Green with white print 312
$32.40USD / Each
MTC Bio   26' Cassette of 12mm Lab Tape, Red w/ White Print
Buy More Save More
EW-06530-56 Mfr # L9010-12RW
COMPARE
Red with white print 312
$32.40USD / Each
MTC Bio   20' Cassette of 12mm Ultra Low Temperature Tape, White w/ Black Print
Buy More Save More
EW-06530-57 Mfr # L9010-12ULT
COMPARE
White with black print 240
$36.30USD / Each
MTC Bio 26' Cassette of 12mm Lab Tape, White w/ Black Print
Buy More Save More
EW-06530-58 Mfr # L9010-12WK
COMPARE
White with black print 312
$38.90USD / Each
MTC Bio   26' Cassette of 12mm Lab Tape, Yellow w/ Black Print
Buy More Save More
EW-06530-59 Mfr # L9010-12YK
COMPARE
Yellow with black print 312
$32.40USD / Each
MTC Bio   26' Cassette of 18mm Lab Tape, White w/ Black Print
Buy More Save More
EW-06530-60 Mfr # L9010-18WK
COMPARE
White with black print 312
$37.60USD / Each
MTC Bio   26' Cassette of 24mm Lab Tape, Clear w/ Black Print
Buy More Save More
EW-06530-61 Mfr # L9010-24CK
COMPARE
Clear with black print 312
$53.20USD / Each
MTC Bio   20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
Buy More Save More
EW-06530-62 Mfr # L9010-24ULT
COMPARE
White with black print 240
$53.20USD / Each
MTC Bio   26' Cassette of 24mm Lab Tape, White w/ Black Print
Buy More Save More
EW-06530-63 Mfr # L9010-24WK
COMPARE
White with black print 312
$48.00USD / Each
MTC Bio   26' Cassette of 6mm Lab Tape, Clear w/ Black Print
Buy More Save More
EW-06530-64 Mfr # L9010-6CK
COMPARE
Clear with black print 312
$36.30USD / Each
MTC Bio   26' Cassette of 6mm Lab Tape, White w/ Black Print
Buy More Save More
EW-06530-65 Mfr # L9010-6WK
COMPARE
White with black print 312
$35.00USD / Each
Showing 1- 13 of 13