Nordson Divergent™ 1-Way Stopcocks with Luer Fittings, Polycarbonate
Easily control the flow of a liquid or gas using an on/off diverter
  • Diverter arrows indicate flow direction
  • Low-torque handle provides smooth adjustments
  • DEHP free
  • ISO 80369-7 compliant
Add a stopcock to your system for an easy on/off control of your fluid. One-way stopcocks feature one female Luer fitting and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Diverter arrows indicate flow direction, enhancing operational efficiency and minimizing errors. Transparent polycarbonate body makes it easy to see the flow.
$3.90 - $109.25USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Luer Connection Type (Port 2) Max Operating Pressure (PSI) Quantity
Availability Pricing
Nordson Divert 1-Way Stopcock, Gamma-Stable Polycarbonate, Female Luer x Male Luer; 1/Ea EW-56140-88 Mfr # FCVFM-001
COMPARE
Male Luer 50 1
$3.90USD / Each
Nordson Divert 1-Way Stopcock, Polycarbonate, Female Luer x Locking Male Luer; 1/Ea EW-56140-89 Mfr # FCVFMR-001
COMPARE
Locking Male Luer 50 1
$4.40USD / Each
Nordson Divert 1-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Locking Male Luer; 1/Ea EW-56140-91 Mfr # FCVHPFMR-001
COMPARE
Locking Male Luer 500 1
$7.20USD / Each
Nordson Divert 1-Way Stopcock, Gamma-Stable Polycarbonate, Female Luer x Male Luer; 25/Pk EW-56140-96 Mfr # FCVFM-001
COMPARE
Male Luer 50 25
$64.10USD / Pkg of 25
Nordson Divert 1-Way Stopcock, Polycarbonate, Female Luer x Locking Male Luer; 25/Pk EW-56140-97 Mfr # FCVFMR-001
COMPARE
Locking Male Luer 50 25
$109.25USD / Pkg of 25
Nordson Divert 1-Way Stopcock, High Pressure, Polycarbonate, Female Luer x Locking Male Luer; 25/Pk EW-56140-99 Mfr # FCVHPFMR-001
COMPARE
Locking Male Luer 500 25
$109.25USD / Pkg of 25