North Sciences/Traceable® TSi Series Ultra-Low Temperature Freezers
The TSi series ultra-low freezers are designed to exceed all standards in reliability, sustainability, and temperature management
  • Cloud-based monitoring provided by TraceableLive® to securely monitor your critical environments and notify your AppleWatch®, smartphone, tablet, or PC if anything goes wrong
  • Audible and visual alarms help those present in the lab identify any out-of-range conditions
  • Ideal for use in healthcare and vaccine monitoring (VFC compliant)
  • Variable speed inverter drive technology, designed to adapt to user patterns, substantially reduces energy and HVAC costs without compromising performance
TSi series ultra-low freezers can get down to -86°C. The hyper insulated cabinet has double injected foam for both inner and outer doors combined with VIP thermal insulation technology. This reduces energy cost and ensures the protection of your samples. Controlling the temperature is easy to with a large 10-inch touch screen oriented for superior user experience. Now you can monitor everything from the inside out, 24/7. The freezers feature an integrated Traceable® data logger and include a one-year Standard subscription plan to TraceableLIVE® cloud-based control and monitoring service.
$12,995.00 - $24,800.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Min Temperature - Freezer (° C) Power (VAC)
Availability Pricing
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 360 L, with TraceableLive®, 120V
Live Image -
EW-44261-69 Mfr # TSI8613-NA
COMPARE
13 -86 120
$15,350.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 360 L, with TraceableLive®, 220V
Live Image -
EW-44261-70 Mfr # TSI8613-EU
COMPARE
13 -86 220
$12,995.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 480 L, with TraceableLive®, 120V
Live Image -
EW-44261-71 Mfr # TSI8617-NA
COMPARE
17 -86 120
$17,125.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 480 L, with TraceableLive®, 220V
Live Image -
EW-44261-72 Mfr # TSI8617-EU
COMPARE
17 -86 220
$14,530.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 590 L, with TraceableLive®, 120V
Live Image -
EW-44261-73 Mfr # TSI8621-NA
COMPARE
21 -86 120
$19,490.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 590 L, with TraceableLive®, 220V
Live Image -
EW-44261-74 Mfr # TSI8621-EU
COMPARE
21 -86 220
$16,535.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 730 L, with TraceableLive®, 120V
Live Image -
EW-44261-75 Mfr # TSI8625-NA
COMPARE
25 -86 120
$22,440.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 730 L, with TraceableLive®, 220V
Live Image -
EW-44261-76 Mfr # TSI8625-EU
COMPARE
25 -86 220
$19,075.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 990 L, with TraceableLive®, 120V
Live Image -
EW-44261-77 Mfr # TSI8630-NA
COMPARE
30 -86 120
$21,080.00USD / Each
North Sciences/Traceable TSi Series ULT Freezer, -40°C to -86°C ULT Freezer, 990 L, with TraceableLive®, 220V
Live Image -
EW-44261-78 Mfr # TSI8630-EU
COMPARE
30 -86 220
$24,800.00USD / Each
Showing 1- 10 of 10