NSI Lab Solutions Oil & Grease, Sulfide, and Surfactant Certified Reference Materials
Validate analytical methods and evaluate the quality of your results with certified reference materials
  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Supplied ready to use—no lengthy preparative steps required
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.
$57.00 - $284.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Packaging Volume (mL)
Availability Pricing
COMPARE
Dodecene-1 LAS 1/EA 10
$77.00USD / Each
COMPARE
Oil, Grease 1/EA 1000
$75.00USD / Each
COMPARE
Oil, Grease 4/PK 1000
$195.00USD / Pkg of 4
COMPARE
Sulfide (from Na2S) 6/PK 2
$284.00USD / Pkg of 6
COMPARE
Sulfide (from Na2S) 1/EA 2
$57.00USD / Each
COMPARE
Sulfide (from Na2S) 6/PK 2
$284.00USD / Pkg of 6
COMPARE
Sulfide (from Na2S) 1/EA 2
$57.00USD / Each
Showing 1- 7 of 7