NSI Lab Solutions Quantitative Hemp Microbiology Proficiency Test Cultures
Ensure state and accreditation regulation compliance with proficiency testing
  • Designed for quantitative determination
  • Two studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Applicable to cultural techniques
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$132.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism type
Availability Pricing
COMPARE
Quantitative APC/TVC in Hemp Proficiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative APC/TVC in Edible Procifiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative BTGN in Hemp Proficiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative BTGN in Edible Procifiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative Coliforms/E. coli in Hemp Proficiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative Coliforms/E. coli in Edible Procifiency Test; 2/PK Bacteria
$132.00USD / Each
COMPARE
Quantitative Yeast/Mold in Edible Procifiency Test; 2/PK Fungi
$132.00USD / Each
COMPARE
Quantitative Yeast/Mold in Hemp Proficiency Test; 2/PK Fungi
$132.00USD / Each
Showing 1- 8 of 8