Ohaus Bead Genie™ Vortex Shaker Accessories
Bead Genie™ shaker accessories to meet your mixing needs
  • Suitable for use with Scientific Industries Bead Genie™ vortex shakers
Choose from a range of tube holders and stainless steel beads for a bead beating solution for any application. Vortex shakers sold separately.
$52.00 - $136.30USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Description
Availability Pricing
Ohaus Bead Genie™ Replacement Top with Screw EW-51402-08 Mfr # SI-B150
COMPARE
Top only Bead Genie™ Replacement Top with Screw
$58.00USD / Each
Ohaus Bead Genie™ Vertical Shaker Holder, 1.5/2.0 mL x 12 Tubes EW-51402-09 Mfr # PP-B100-401
COMPARE
1.5/2.0 mL x 12 Tubes Bead Genie™ Vertical Shaker Holder, 1.5/2.0 mL x 12 Tubes
$52.00USD / Each
Ohaus Bead Genie™ Vertical Shaker Holder, 15 mL x 6 Tubes EW-51402-11 Mfr # PP-B100-402
COMPARE
15 mL x 6 Tubes Bead Genie™ Vertical Shaker Holder, 15 mL x 6 Tubes
$52.00USD / Each
Ohaus Bead Genie™ Vertical Shaker Holder, 50 mL x 3 Tubes EW-51402-12 Mfr # PP-B100-403
COMPARE
50 mL x 3 Tubes Bead Genie™ Vertical Shaker Holder, 50 mL x 3 Tubes
$52.00USD / Each
Ohaus Bead Genie™ Horizontal Shaker Holder, 1.5/2.0 mL x 12 Tubes EW-51402-13 Mfr # PP-B100-404
COMPARE
1.5/2.0 mL x 12 Tubes Bead Genie™ Horizontal Shaker Holder, 1.5/2.0 mL x 12 Tubes
$52.00USD / Each
Ohaus Stainless Steel Beads, 2.4 mm; 100/PK EW-51402-14 Mfr # SI-BS02
COMPARE
2.4 mm diameter bead Stainless Steel Beads, 2.4 mm; 100/PK
$55.30USD / Each
Ohaus Stainless Steel Beads, 3.2 mm; 100/PK EW-51402-16 Mfr # SI-BS03
COMPARE
3.2 mm diameter bead Stainless Steel Beads, 3.2 mm; 100/PK
$55.30USD / Each
Ohaus Stainless Steel Beads, 5 mm; 100/PK EW-51402-17 Mfr # SI-BS05
COMPARE
5 mm diameter bead Stainless Steel Beads, 5 mm; 100/PK
$55.30USD / Each
Ohaus Bead Genie™ Horizontal Plastic Clip Microtube Holder; 24 tubes EW-51402-19 Mfr # SI-HP524
COMPARE
24 tubes Bead Genie™ Horizontal Plastic Clip Microtube Holder; 24 tubes
$136.30USD / Each
Showing 1- 9 of 9