Ohaus Endeavor™ 5000 Series Orbital Digital Shakers
Triple eccentric drive provides precise, reliable sample mixing
  • Remote function permits PC control and data transfer
  • Bright, backlit LCD display clearly shows speed and time
  • Durable chemical-resistant polymer housing
  • Easy-to-clean aluminum standard tray
  • IP21 ingress protection
These light duty orbital digital shakers provide versatility for different shaking requirements. They are compatible with use in both incubators and cold rooms, ensuring flexibility across a wide range of environmental conditions. Applications include ELISA assays, cell cultures, protein studies, bacterial and yeast cultures, and DNA studies.
The compact design of the shakers saves you valuable bench space, while the advanced microprocessor control optimizes speed ramp-up to avoid splashing. The triple eccentric drive and maintenance-free brushless DC motor offers stable, accurate, and long-term shaking. A simple on-board menu allows you to set, store, and recall multi-step programs, while the RS232 interface provides two-way communication for data logging and remote control of the unit.
Safety features include an integrated overload and unbalanced detection to protect samples from potential damage, ensuring the reliability of experimental results. A visual and audio alarm system alerts you when the timer counts down to zero or an error occurs.
The easy-to-clean aluminum tray features 11 mounting points for various accessories allowing you to customize your shaker with tube racks, flask clamps, and a non-slip rubber mat.
The shakers are available in three models: 3 mm orbit for gentle mixing, 20 mm orbit for more vigorous applications, and the dedicated microplate model.
$998.80 - $1,399.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Orbit Size (mm)
Availability Pricing
Ohaus E51LD0403 Endeavor Standard Duty Orbital Digital Shaker with 3mm Orbit, 100 to 800 rpm EW-53026-81 Mfr # E51LD0403
COMPARE
800 100 3
$1,399.00USD / Each
Ohaus E51LD0420 Endeavor Standard Duty Orbital Digital Shaker with 20mm Orbit, 50 to 300 rpm EW-53026-82 Mfr # E51LD0420
COMPARE
300 50 20
$1,299.00USD / Each
Ohaus E51LDMP03 Endeavor Standard Duty Orbital Digital Microplate Shaker with 3mm Orbit, 100 to 1200 rpm EW-53026-83 Mfr # E51LDMP03
COMPARE
1200 100 3
$998.80USD / Each